Skip to Content
Merck
All Photos(2)

Key Documents

SYMBECS01

Millipore

Steritest® ampoule breaker end cap with seal

This accessory streamlines the workflow and increases safety during sterility testing

Synonym(s):

Lid with gasket

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€447.00

€447.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 EACH
€447.00

About This Item

UNSPSC Code:
23151818
NACRES:
NB.51

€447.00


Please contact Customer Service for Availability

Request more information

Agency

EP (2.6.1)
JP (4.06)
USP (71)

manufacturer/tradename

Steritest®

input

pharmaceutical(s)

application(s)

pharmaceutical
sterility testing

compatibility

for use with Steritest® Symbio pump

shipped in

ambient

General description

Steritest® ampoule breaker replacement kit containing end cap with seal.

Application

Accessories for Streamlining Your Workflow and Increasing Safety during sterility testing.

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service