Skip to Content
Merck
All Photos(1)

Key Documents

SCT038

Sigma-Aldrich

BioTracker Orange OH and HClO Dye

Live cell imaging probe to detect hydroxy radical (OH) or hypochlorous acid (HClO) in live cells.

Sign Into View Organizational & Contract Pricing

Select a Size

SCT038
€593.00

€593.00


Estimated to ship onMay 30, 2025Details



Select a Size

Change View
SCT038
€593.00

About This Item

UNSPSC Code:
12352207
NACRES:
NA.47

€593.00


Estimated to ship onMay 30, 2025Details


form

lyophilized

application(s)

cell analysis

detection method

fluorometric

General description

Reactive oxygen species (ROS) are important mediators in a variety of biological and pathological events. For example, ROS generated in mitochondria play important roles in the modulation of signal transduction cascades and transcription factors. However, oxidative stress due to excessive generation of ROS is implicated in many human diseases, including acute and chronic inflammatory processes, ischemic stroke, diabetes, sepsis, ischemia-reperfusion injury, atherosclerosis, and neurodegenerative disorders.BioTracker Orange OH and HClO Dye is an orange fluorescent probe to detect hydroxy radical (・OH) or hypochlorous acid (HClO) in live-cell imaging. It can detect -OH and HClO among other reactive oxygen species (O2-, H2O2, NO, ONOO-). Its nearly red fluorescence spectrum allows multicolor imaging with green (ex. GFP, FITC) and blue (ex. Hoechst 33342) fluorophores. Because of its positive charge, OxiORANGE tends to localize within mitochondria. It has high photostability and is suitable for time-lapse imaging of intracellular hROS generation.Spectral Properties:Absorbance (max): 553mEmission (max): 577nmResearch Category:Live Cell Dye

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service