Skip to Content
Merck
All Photos(3)

Key Documents

RDYSTRM80

Millipore

ReadyStream® Barcode Reader

Barcode reader to be used with the ReadyStream® system for inputting data matrix codes

Synonym(s):

Barcode Reader, Scanner

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€298.00

€298.00


Estimated to ship onFebruary 17, 2025Details


Request more information

Select a Size

Change View
1 EACH
€298.00

About This Item

UNSPSC Code:
13102014
NACRES:
NB.51

€298.00


Estimated to ship onFebruary 17, 2025Details


Request more information

packaging

pkg of 1 unit

system L × W × H

26 cm × 21 cm × 9 cm

application(s)

food and beverages

compatibility

for use with ReadyStream® System

storage temp.

15-25°C

General description

ReadyStream® Barcode Reader is used to conveniently scan and register information from the ReadyStream® Media Bags and Filter Sets as well as to set pre-defined testing parameters in the system by scanning your custom data matrix code generated with the web tool.

Application

ReadyStream® Barcode Reader is designed to be used with ReadyStream® System. The bar codes assigned to the systems enhance data protection, transparency, and traceability.

Other Notes

To access the ReadyStream® Data Matrix Generator Tool follow this link https://www.sigmaaldrich.com/US/en/support/calculators-and-apps/readystream-data-matrix-generator

Legal Information

READYSTREAM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service