Recommended Products
grade
for protein analysis
Quality Level
description
ED<sub>80</sub> = 93 ± 3 pg/mL ED<sub>50</sub> = 230 ± 15 pg/mL ED<sub>20</sub> = 558 ± 44 pg/mL
analytes available: PYY
20-1280 pg/mL (standard curve range)
feature
accuracy 94.5 ± 10.2%
species reactivity
human
sensitivity measuring range
20 pg/mL
technique(s)
radioimmunoassay: suitable
UniProt accession no.
detection method
radioactive
General description
SIZE: 97 amino acids; 11046 Da
SUBCELLULAR LOCATION: Secreted.
SIMILARITY: SwissProt: P10082 ## Belongs to the NPY family.
Specificity
Application
Analysis Note
Disclaimer
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 1B
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service