PCC300C30E
Pellicon® Capsule with Ultracel® Membrane
C Screen, 3 m2 manifold extender AseptiQuik® L connectors
Synonym(s):
Pellicon® Capsule with Ultracel® Membrane
About This Item
Recommended Products
material
EPDM rubber seal
PPO/PS blend (core
core)
PPO/PS blend housing
TPE seal
composite regenerated cellulose (CRC) membrane
epoxy adhesive
polyester screen
polypropylene seal
polyurethane adhesive
Quality Level
sterility
sterile; γ-irradiated
product line
Pellicon®
feature
holdup volume 1290 mL (in feed channel)
holdup volume 1872 mL (in permeate channel)
packaging
package of 1
parameter
max. inlet pressure at 60 psi (4.1 bar) and 4-30 °C
max. transmembrane pressure at 45 psi (3.1 bar) and 4-30 °C
4-6 L/min-m2 flow rate
technique(s)
ultrafiltration: suitable
filtration area
3 m2
screen size
, Type C screen (coarse screen)
size
45.9 cm (18.1 in.) , Length
46.9 cm (18.5 in.) , Height
width
48.2 cm (19.0 in.)
482 mm
impurities
≤5 ppm Meets TOC (in retentate effluent, after a WFI flush of 20 L/m²)
matrix
Ultracel®
pore size
300 kDa
pH range
2-13
storage temp.
15-25°C
Looking for similar products? Visit Product Comparison Guide
General description
Permeate: AseptiQuik® G Connector
Application
Features and Benefits
- Plug ′n play, holderless design - easy to install, safe to remove
- Gamma Sterilized and preseravtive-free - ready to process in minutes
- True single-use, self-contained capusle - fast, safe, and flexible batch turnaround
- Proven Ultracel® membrane and C screen - high recovery, superior mass transfer, solvent resistance
- Pellicon® TFF proven performance - true linear scalability within Pellicon® TFF families
Packaging
Other Notes
- Comprehensive documentation to support qualification, risk assessment and process optimization needs.
- Consolidation of product specific testing, quality and regulatory information to simplify compliance requirements.
- Convenient 24/7 access to up-to-date product information.
Legal Information
related product
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Our overview of scalable tangential flow filtration for viral vector production will provide key considerations for developing and optimizing your downstream process.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service