Skip to Content
Merck
All Photos(1)

Key Documents

P2PLGCC01

Millipore

Pellicon® 2 Cassette Filter

NMWCO 10 kDa

Synonym(s):

Pellicon 2 47 0.1 μm

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€1,690.00

€1,690.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
€1,690.00

About This Item

UNSPSC Code:
23151806

€1,690.00


Please contact Customer Service for Availability

material

regenerated cellulose

Quality Level

manufacturer/tradename

Pellicon®

technique(s)

analytical sample preparation: suitable
protein purification: suitable

membrane nominal area

0.1 m2

gravimetric extractables

≤1500 mg/m2

matrix

Biomax®

pore size

10 kDa NMWCO

Looking for similar products? Visit Product Comparison Guide

General description

Device Configuration: Mini Cassette

Features and Benefits

Type C screen (coarse screen)

Other Notes

These products are manufactured in a facility which adheres to FDA Good Manufacturing Practices.

Analysis Note

Gravimetric Extractables: after a water flush

Legal Information

BIOMAX is a registered trademark of Merck KGaA, Darmstadt, Germany
Pellicon is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service