Skip to Content
Merck
All Photos(1)

Key Documents

P0DC0VER1

Milli-Q

POD Protection in Silicone Crystal

Protects the POD base against repeated exposure to reactive substances such as acid vapours

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€453.00

€453.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 EA
€453.00

About This Item

UNSPSC Code:
41104200
eCl@ss:
33050190
NACRES:
JA.14

€453.00


Please contact Customer Service for Availability

Request more information

packaging

pkg of 1 unit

compatibility

for use with E-POD®

shipped in

ambient

General description

Silicone crystal protects the POD (Point of Dispense) base against repeated exposure to reactive substances, such as acid vapors, harsh chemicals like strong acid and bases, aggressive solvents or etchants.

Legal Information

E-POD is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service