Skip to Content
Merck
All Photos(1)

Key Documents

MXQP0WEU1

Millipore

Power Supply for Milliflex® Quantum Reader

Milliflex® Quantum Rapid Detection System power supply for European countries

Synonym(s):

EU Power Supply, For rapid detection of microbial contamination in filterable samples, suitable for bioburden testing

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€144.00

€144.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 EA
€144.00

About This Item

UNSPSC Code:
41116129
NACRES:
NB.79

€144.00


Please contact Customer Service for Availability

Request more information

manufacturer/tradename

Milliflex®

compatibility

for use with Milliflex® Quantum Reader

Looking for similar products? Visit Product Comparison Guide

General description

EU Power Supply for Milliflex® Quantum Reader is designed to provide constant voltage/current to avoid process fluctuation.

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service