Skip to Content
Merck
All Photos(1)

Key Documents

MSVMHTS06

Millipore

On/Off Valve Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€275.00

€275.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
€275.00

About This Item

UNSPSC Code:
41104923
eCl@ss:
32039006
NACRES:
NB.22

€275.00


Please contact Customer Service for Availability

product line

MultiScreen®

Quality Level

manufacturer/tradename

MultiScreen®

shipped in

ambient

General description

MultiScreen vacuum manifolds have been developed for all vacuum filtration applications using MultiScreenHTS and MultiScreen filter plates. Manifold components are resistant to solvents and suitable for use with automated or manual protocols.

New MultiScreenHTS Vacuum Manifold with DirectStack technology

The new MultiScreenHTS vacuum manifold provides enhanced performance for use with MultiScreenHTS 96- and 384-well filter plates as well as the Solvinert family of filter plates. It can be easily configured for both flow-to-waste and analyte collection. For applications that require filtrate collection for further analysis, the MultiScreenHTS vacuum manifold and filter plates offer a significant improvement in reliability over current systems. When used with MultiScreenHTS filter plates and ANSI/SBS standard receiver plates, the unique plate and manifold designs combine to eliminate gaps between the flow directors and receiver wells and provide a plate-on-plate processing option for assays where filtrate collection is required. Direct stacking also makes vacuum initiations effortless. Complete filtration assay cycles can be done with no manual intervention.

The MultiScreenHTS vacuum manifold features:
- DirectStack technology for crosstalk-free filtrate collection
- Configurations for deep well or standard receiver plates
- ANSI/SBS compliant footprint allows for easy robotic deck integration


Also available: MultiScreen Vacuum Manifold for classic filter plates.

Chemical Compatibility
- The compatibility list, which you can find on General Guidelines for MultiScreen Plates covers chemicals which are compatible with the MultiScreenHTS and MultiScreen vacuum manifolds, as well as MultiScreen Solvinert filter plates.

Applications
-Vacuum Filtration of Multiwell Filter Plates, Automated Liquid Handling

Legal Information

MULTISCREEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service