Skip to Content
Merck
All Photos(1)

Key Documents

MSP010054

Millipore

Milliflex Pre-filled Agar Cassettes (R2A)

suitable (for bioburden testing of potable water)

Synonym(s):

Reasoner’s 2A agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NA.74
Product MSP010054 is not currently sold in your country. Contact Technical Service

description

General microbiological analysis

Quality Level

sterility

sterile

form

plate (pre-poured agar)

manufacturer/tradename

Milliflex®

input

sample type pharmaceutical(s)

pH

7.2±0.2

suitability

suitable (for bioburden testing of potable water)

application(s)

bioburden testing
cosmetics
microbiology
pharmaceutical
water monitoring

shipped in

wet ice

storage temp.

2-8°C

General description

R2A agar was devised by Reasoner and Geldreich for the isolation and cultivation of bacteria from treated potable water. The medium is suitable for this purpose as it is not nutrient-rich, and at lower temperatures and with longer incubation times it stimulates the growth of stress and chlorine-tolerant bacteria. These would normally be inhibited when cultured on nutrient-rich media by faster-growing microbes. The agar-based media comes in ready-to-use cassettes Each cassette is a plastic 3-piece assembly containing approximately 10ml of R2A Agar.

Application

R2A agar is used in heterotrophic plate count (HPC) testing. HPC, formerly known as the standard plate count, is a procedure for estimating the number of live bacteria in water and measuring changes during water treatment in distribution systems or in swimming pools.

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service