Key Documents
MAVFLTVP2
Millipore
MAS-100 VF® Portable Air Sampler Validation Protocol
for use with MAS-100 VF® Microbial Air Samplers
About This Item
UNSPSC Code:
41116500
NACRES:
JA.11
Product MAVFLTVP2 is not currently sold in your country. Contact Technical Service
availability
available only in North America
compatibility
for use with MAS-100 VF® Microbial Air Samplers
General description
MAS-100 VF® Validation Protocol(Letter Format) is designed as a ready to use step by step guide to perform IQ and OQ activities of MAS-100 VF® microbial air sampling systems. The protocol contains an exhaustive list of tests to cover a batch of units to be validated, the test record protocol MAVFLTTR1 including test descriptions and test records must be ordered separately. The protocol follows GMP, EP, USP and JP regulatory guidelines. Only available in North America.
Application
MAS-100 VF® Validation Protocol is used to validate microbial air sampling system with ease and consistent performance while following regulatory guidelines.
Legal Information
MAS-100 VF is a registered trademark of MBV AG