Skip to Content
Merck
All Photos(1)

Key Documents

MABS2267

Sigma-Aldrich

Anti-ANT1/SLC25A4 Antibody, clone IF3F11

Synonym(s):

ADP/ATP carrier protein 1, ADP/ATP carrier protein, heart/skeletal muscle isoform T1, ADP/ATP translocase 1, Adenine nucleotide translocator 1, Solute carrier family 25 member 4

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€138.00
100 μL
€348.00

€138.00


Estimated to ship onMarch 31, 2025Details


Request a Bulk Order

Select a Size

Change View
25 μL
€138.00
100 μL
€348.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

€138.00


Estimated to ship onMarch 31, 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

IF3F11, monoclonal

mol wt

calculated mol wt 33.06 kDa
observed mol wt ~33 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100

technique(s)

western blot: suitable

isotype

IgG1κ

epitope sequence

Internal

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

Gene Information

human ... SLC25A4(291)

Specificity

Clone IF3F11 is a mouse monoclonal antibody that detects Adenine nucleotide translocator 1 (ANT1/SLC25A4). It targets an epitope within 15 amino acids from the internal region.

Immunogen

His-tagged, full-length, recombinant human ADP/ATP translocase 1 (ANT1).

Application

Quality Control Testing

Evaluated by Western Blotting in Human heart tissue lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected ANT1/SLC25A4 in Human heart tissue lysate.

Tested Applications

Western Blotting Analysis: A 1:500 dilution from a representative lot detected ANT1/SLC25A4 in Human skeletal muscle and Human brain tissue extracts.

Western Blotting Analysis: A representative lot detected ANT1/SLC25A4 in Western Blotting applications (Lu, Y-W., et al. (2017). Mol Biol Cell. 28(11):1489-1500; Acoba, M.G., et al. (2021). Cell Rep. 34(11):108869).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

ADP/ATP translocase 1 (UniProt: P12235; also known as ADP/ATP carrier protein 1, ADP/ATP carrier protein, heart/skeletal muscle isoform T1 1 , Adenine nucleotide translocator 1, ANT 1, Solute carrier family 25 member 4) is encoded by the SLC25A4 (also known as AAC1, ANT1) gene (Gene ID: 291) in human. ANT1 is a multi-pass inner mitochondrial membrane protein with six transmembrane domains. It serves as an ADP:ATP antiporter that mediates import of ADP into the mitochondrial matrix for ATP synthesis, and export of ATP out to provide energy for cellular functions. It is reported to cycle between the cytoplasmic-open state (c-state) and the matrix-open state (m-state) and operates by the alternating access mechanism with a single substrate-binding site intermittently exposed to either the cytosolic or matrix side of the inner mitochondrial membrane. It is also involved in mitochondrial uncoupling and mitochondrial permeability transition pore activity. ANT also acts as a regulator of mitophagy independently of ADP:ATP antiporter activity. It promotes mitophagy via interaction with TIMM44 that leads to inhibition of presequence translocase TIMM23, thereby promoting stabilization of PINK1. ANT1 deficiency has been implicated in various pathological states, including hypertrophic cardiomyopathy, mitochondrial myopathy, lactic acidosis, and progressive external opthalmoplegia. (Ref.: Hoshino, A., et al. (2019). Nature. 575(7782); 375-379; Lu, Y-W., et al. (2017). Mol. Biol. Cell. 28(11):1489-1506; Kawamaoto, H., et al. (2011). Hum. Mol. Genet. 20(15); 2964-2974).

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Reconstitution

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service