Skip to Content
Merck
All Photos(1)

Key Documents

MABS2174

Sigma-Aldrich

Anti-PNPLA3 Antibody, clone 11C5

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€142.00
100 μG
€363.00

€142.00


Estimated to ship onMarch 31, 2025Details


Request a Bulk Order

Select a Size

Change View
25 μG
€142.00
100 μG
€363.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€142.00


Estimated to ship onMarch 31, 2025Details


Request a Bulk Order

antibody form

purified antibody

clone

11C5
monoclonal

purified by

using protein G

species reactivity

mouse, human, human

concentration

(Please refer to lot specific datasheet.)

technique(s)

western blot: suitable

isotype

IgG2aκ

UniProt accession no.

General description

Recombinant fusion protein of human PNPLA3 and Trigger Factor.

Specificity

Clone 11C5 is a mouse monoclonal antibody that detects human Patatin-like phospholipase domain-containing protein 3 (PNPLA3).

Application

Western Blotting Analysis: A representative lot detected PNPLA3 in Western Blotting applications (BasuRay, S., et. al. (2019). Proc. Natl. Acad. Sci. USA. 116(19):9521-9526).

Quality

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected PNPLA3 in HepG2 cell lysate.

Physical properties

~54 kDa observed; 52.86 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service