Skip to Content
Merck
All Photos(1)

Key Documents

MABS1300

Sigma-Aldrich

Anti-asparagine synthetase Antibody, clone Z5808

clone Z5808, from mouse

Synonym(s):

Asparagine synthetase [glutamine-hydrolyzing], Cell cycle control protein TS11, Glutamine-dependent asparagine synthetase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€365.00

€365.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€365.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€365.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

Z5808, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... ASNS(440)

General description

Asparagine synthetase [glutamine-hydrolyzing] (UniProt: P08243; also known as EC:6.3.5.4, Cell cycle control protein TS11, Glutamine-dependent asparagine synthetase) is encoded by the ASNS (also known as TS11) gene (Gene ID: 440) in human. Asparagine synthetase catalyzes the biosynthesis of L-asparagine from L-aspartate in an ATP-dependent manner. It contains one asparagine synthetase domain and one glutamine amidotransferase type-2 domain. ASNS transcription is enhanced by amino acid deprivation via a signal transduction pathway that senses amino acid availability. Mutations in ASNS gene are reported to result in asparagine synthetase deficiency that is characterized by microcephaly, severely delayed psychomotor development, progressive encephalopathy, and hyperekplexic activity. There has been much interest in the development of asparagine synthetase inhibitors for treatment of asparaginase-resistance leukemias and levels of asparagine synthetase have been considered as a potential biomarker in ovarian cancers.

Specificity

Clone Z5808 detects Asparagine synthetase in human creast cancer, human tonsil, and human uterus. It targets an epitope within 50 amino acids from the N-terminal end.

Immunogen

N-asparagine synthetase-budded virus.

Application

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected asparagine synthetase in human tonsil and human uterus tissues.

Immunocytochemistry Analysis: A representative lot detected asparagine synthetase in Immunocytochemistry applications (Kusano-Arai, O., et. al. (2012). Hybridoma (Larchmt). 31(5):325-32).

Flow Cytometry Analysis: A representative lot detected asparagine synthetase in Flow Cytometry applications (Kusano-Arai, O., et. al. (2012). Hybridoma (Larchmt). 31(5):325-32).

ELISA Analysis: A representative lot detected asparagine synthetase in ELISA applications (Kusano-Arai, O., et. al. (2012). Hybridoma (Larchmt). 31(5):325-32).

Western Blotting Analysis: A representative lot detected asparagine synthetase in Western Blotting applications (Kusano-Arai, O., et. al. (2012). Hybridoma (Larchmt). 31(5):325-32).
This mouse monoclonal Anti-asparagine synthetase, clone Z5808, Cat. No. MABS1300 has been tested for use in ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting for the detection of human Asparagine Synthetase.

Quality

Evaluated by Immunohistochemistry in human breast cancer tissues.

Immunohistochemistry Analysis: A 1:50 dilution of this antibody detected asparagine synthetase in human breast cancer tissue.

Target description

64.37 kDa calculated.

Physical form

Format: Purified
Purified mouse monoclonal antibody IgG2a in PBS with 0.01% sodium azide.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service