Skip to Content
Merck
All Photos(2)

Key Documents

MABS1228

Sigma-Aldrich

Anti-ANKRD1/CARP Antibody, clone 12G8.2

clone 12G8.2, from mouse

Synonym(s):

Ankyrin repeat domain-containing protein 1, Cardiac ankyrin repeat protein, Cytokine-inducible gene C-193 protein, Cytokine-inducible nuclear protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€368.00

€368.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€368.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€368.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

12G8.2, monoclonal

species reactivity

rat, human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ANKRD1(27063)

General description

Ankyrin repeat domain-containing protein 1 (UniProt Q15327; also known as Cardiac ankyrin repeat protein, Cytokine-inducible gene C-193 protein, Cytokine-inducible nuclear protein) is encoded by the ANKRD1 (also known as C193, CARP, HA1A2) gene (Gene ID 27063) in human. ANKRD1/CARP belongs to a family of conserved muscle ankyrin repeat proteins (MARPs), members of which also include ANKRD2 and diabetes ankyrin repeat protein (DARP, ANKRD23). ANKRD1 is highly expressed in cardiomyocytes and known to interact with myopalladin and titin, implicating its roles in myopalladin complex-dependent sarcomere integrity maintenance and stress-induced gene regulations medidated by the titin complex. ANKRD1 is up-regulated in patients with dilated cardiomyopathy and in animal models of cardiac hypertrophy induced by pressure overload and adrenergic stimulation. ANKRD1 is shown to regulate the hypertrophic agonist phenylephrine-/PE-induced α-adrenergic signaling pathway in neonatal rat ventricular myocytes, where it recruits and localizes ERK1/2 and GATA4 in a sarcomeric complex, resulting in enhanced GATA4 phosphorylation and subsequent ANKRD1 complex nuclear translocation and gene activation.

Immunogen

Epitope: Ankyrin repeats 4&5.
GST-tagged recombinant human ANKRD1/CARP fragment corresponding ankyrin repeats 4&5.

Application

Anti-ANKRD1/CARP Antibody, clone 12G8.2 is an antibody against ANKRD1/CARP for use in Western Blotting, Immunohistochemistry (Paraffin).
Immunohistochemistry Analysis: A 1:250-1,000 dilution from a representative lot detected ANKRD1/CARP in human and rat heart tissue sections.

Quality

Evaluated by Western Blotting in human heart tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected ANKRD1/CARP in 10 µg of human heart tissue lysate.

Target description

~37 kDa observed. 36.25 kDa calculated. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lynn Yap et al.
NPJ Regenerative medicine, 8(1), 26-26 (2023-05-27)
Ischemic heart disease, which is often associated with irreversibly damaged heart muscle, is a major global health burden. Here, we report the potential of stem cell-derived committed cardiac progenitors (CCPs) have in regenerative cardiology. Human pluripotent embryonic stem cells were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service