Skip to Content
Merck
All Photos(1)

Key Documents

MABS1196

Sigma-Aldrich

Anti-SENP1 Antibody, clone 8B6.1

culture supernatant, clone 8B6.1, from mouse

Synonym(s):

Sentrin-specific protease 1, Sentrin/SUMO-specific protease SENP1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€313.00

€313.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
€313.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€313.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

8B6.1, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG2bλ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SENP1(29843)

General description

Sentrin-specific protease 1 (EC 3.4.22.68; UniProt Q9P0U3; also known as Sentrin/SUMO-specific protease SENP1) is encoded by the SENP1 gene (Gene ID 29843) in human. SUMOylation, protein posttranslation modification by small ubiquitin-like modifier (SUMO), is a signaling event in many cellular processes. As is the case with ubiquitination, SUMOylation is a reversible process. SUMO E1 activating enzyme, E2 conjugating enzyme, and E3 ligase mediate SUMOylation of substrate proteins, while a family of SUMO-specific proteases (SENPs) are responsible for the deSUMOylation. Seven SENPs are identified in human (SENP1-3 and SENP5-8). SENP1 processes full-length SUMO1, SUMO2 and SUMO3 to their mature forms and mediates their removal (deconjugation) from substrate proteins, including BHLHE40/DEC1, CCAR2, CLOCK, DBC1, HADC1, HIPK2, MTA1, p53, and SIRT1. SENP1 is reported to be overexpressed and induced by IL-6 in multiple myeloma (MM) cells, and SENP1-knockdown triggers apoptosis and reduces viability, proliferation and colony forming ability of MM cells.

Specificity

Expected to react with both spliced isoforms of human SENP1 reported by UniProt (Q9P0U3).

Immunogen

Epitope: Internal (C-terminal half).
GST-tagged recombinant fragment from the C-terminal half of human SENP1.

Application

Anti-SENP1 Antibody, clone 8B6.1 is an antibody against SENP1 for use in Western Blotting.
Research Category
Signaling
Research Sub Category
General Post-translation Modification

Quality

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: A 1:2,000 dilution of this antibody detected SENP1 in 10 µg of HepG2 cell lysate.

Target description

~73 kDa observed. 73.48 kDa (isoform 1), 73.35 kDa (isoform 2) calculated.

Physical form

Mouse monoclonal IgG2bλ hybridoma culture supernatant with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service