Skip to Content
Merck
All Photos(2)

Key Documents

MABN873

Sigma-Aldrich

Anti-Kallikrein 6/KLK6 Antibody, clone E24

clone E24, from mouse

Synonym(s):

Kallikrein-6, hK6, Neurosin, Protease M, Serine protease 18, Serine protease 9, SP59, Zyme

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€368.00

€368.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€368.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€368.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

E24, monoclonal

species reactivity

mouse, human

technique(s)

ELISA: suitable
affinity chromatography: suitable
immunohistochemistry: suitable (paraffin)

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KLK6(5653)

General description

Kallikrein-6 (UniProt Q92876; also known as hK6, Neurosin, Protease M, Serine protease 18, Serine protease 9, SP59, Zyme) is encoded by the KLK6 (also known as PRSS18, PRSS9) gene (Gene ID 5653) in human. Kallikrein-6 belongs to the kallikrein subfamily of serine proteases, members of which are involved in diverse physiological functions and many are used clinically as biomarkers for cancers. Originally identified due to its aberrant expression in mammary and ovarian cancers, kallikrein-6 is expressed in a wide range of tissues, including breast, kidney, and the central nervous system. Kallikrein-6 is known to cleave components of the extracellular matrix (ECM) and is reported to be involved in multiple cellular processes, including inflammation, receptor activation, and apoptosis. In addition, deregulated KLK6 expression occurs in neurodegenerative and skin disorders, and is a common event in many types of human cancers. Consequently, KLK6 represents a promising biomarker of early diagnosis and/or unfavorable prognosis in these human malignancies. Kallikrein-6 is initially produced as a preproprotein with a signal (aa 1-16) and a propeptide (aa 17-21), cleavage of which releases the mature protein (aa 22-244).

Specificity

Clone E24 reacts with kallikrein-6, but not other kallikrein family members. Reactivity against kallikrein-6 spliced isoforms 2&3 reported by UniProt (Q92876) has not been determined.

Immunogen

Recombinant full-length human Kallikrein 6.

Application

Anti-Kallikrein 6/KLK6 Antibody, clone E24 is an antibody against Kallikrein 6/KLK6 for use in Immunohistochemistry (Paraffin), ELISA, Inhibits Activity/Function, Immunoaffinity Purification.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Kallikrein 6/KLK6 in mouse cerebral cortex tissue.
ELISA Analysis: Representative lots were conjugated with biotin and used as the detection antibody for analyzing kallikrein-6 levels in ovarian cytosolic extracts as well as ascites fluids from ovarian cancer patients (Shan, S.J., et al. (2007). Br. J. Cancer. 96(2):362-372; Luo, L.Y., et al. (2006). Tumor Biol. 27:227-234).
Activity/Function Inhibition Analysis: Representative lots inhibited the catalytic activity of purified human kallikrein-6 (hK6) from ascites fluid as well as recombinant hK6 in cell-free enzymatic assays (Sotiropoulou, G., et al. (2012). Chem. Cent. J. 6(1):148; Luo, L.Y., et al. (2006). Tumor Biol. 27:227-234; Ghosh, M.C., et al. Tumor Biol. (2004). 25(4):193-199).
Immunoaffinity Purification: Representative lots were conjugated to resins and employed for affinity purification of kallikrein-6 from human ascites fluid, cerebrospinal fluid (CSF), and breast milk (Luo, L.Y., et al. (2006). Tumor Biol. 27:227-234; Hutchinson, S., et al. (2003). Clin. Chem. 49(5):746-751).
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Quality

Evaluated by Immunohistochemistry in human cerebral cortex tissue.

Immunohistochemistry Analysis: A 1:50 dilution of this antibody detected Kallikrein 6/KLK6 in human cerebral cortex tissue.

Target description

26.85 kDa calculated.

Physical form

Format: Purified
Purified mouse monoclonal IgG1κ antibody in 0.1 M Sodium bicarbonate buffer, pH 8.2 with 0.02% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service