Skip to Content
Merck
All Photos(3)

Key Documents

MABN814

Sigma-Aldrich

Anti-Twinkle Antibody, clone 13A10.1

clone 13A10.1, from mouse

Synonym(s):

Twinkle protein, mitochondrial, Progressive external ophthalmoplegia 1 protein, T7 gp4-like protein with intramitochondrial nucleoid localization, T7-like mitochondrial DNA helicase, Twinkle

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€381.00

€381.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€381.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€381.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

13A10.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TWNK(56652)

General description

Twinkle protein, mitochondrial (EC 3.6.4.12; UniProt Q96RR1; also known as Ataxin 8, Mitochondrial twinkle protein, Progressive external ophthalmoplegia 1 protein, T7 gp4-like protein with intramitochondrial nucleoid localization, T7 helicase-related protein with intramitochondrial nucleoid localization, T7-like mitochondrial DNA helicase) is encoded by the EPO1 (also known as ATXN8, C10orf2, IOSCA, MTDPS7, PEO, PEO1, PEOA3, PRLTS5, SANDO, SCA8, TWINL) gene (Gene ID 56662) in human. Mitochondrial DNA (mtDNA) replication involves the catalytic and accessory subunits of DNA polymerase gamma, the mtDNA helicase Twinkle, the mitochondrial single-stranded DNA-binding protein, and the mitochondrial RNA polymerase. Cells expressing Twinkle mutants accumulate double-stranded mtDNA replication intermediates with loss of RNA associated with the lagging strand, consistent with the role of Twinkle in unwinding double-stranded mtDNA. Mutations in the human Twinkle gene (C10orf2) cause autosomal dominant progressive external ophthalmoplegia (PEO), associated with multiple mtDNA deletions.

Immunogen

GST-tagged recombinant protein corresponding to the C-terminal of human Twinkle.

Application

This Anti-Twinkle Antibody, clone 13A10.1 is validated for use in Western Blotting, Immunohistochemistry (Paraffin) for the detection of Twinkle.
Western Blotting Analysis: 0.5 µg/mL from a representative lot detected Twinkle in 10 µg of K562 cell lysate.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Twinkle in human cerebral cortex and human skeletal muscle tissue.

Quality

Evaluated by Western Blotting in HL-60 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Twinkle in 10 µg of HL-60 cell lysate.

Target description

~77 kDa observed

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service