Skip to Content
Merck
All Photos(2)

Documents

MABF238

Sigma-Aldrich

Anti-CD38 Antibody, clone 2H3-2

clone 2H3-2, from mouse

Synonym(s):

ADP-ribosyl cyclase 1, Cyclic ADP-ribose hydrolase 1, cADPr hydrolase 1, T10, CD38

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2H3-2, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable
western blot: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CD38(952)

General description

CD38 is also called ADP-ribosyl cyclase 1, Cyclic ADP-ribose hydrolase 1 (cADPr hydrolase 1), and T10. CD38 synthesizes cyclic ADP-ribose posseses cADPr hydrolase activity, and also acts as a receptor in cells of the immune system. CD38 is expressed in pancreas, liver, kidney, brain, testis, ovary, placenta, malignant lymphoma, and neuroblastoma, as well as in both early and late stages of the B and T-cell maturation, and erythroid and myeloid progenitors in bone marrow.

Immunogen

Recombinant protein corresponding to human CD38.

Application

Detect CD38 using this mouse monoclonal antibody, Anti-CD38 Antibody, clone 2H3-2 validated for use in western blotting & Flow Cytometry.
Flow Cytometry Analysis: 1 µg from a representative lot detected CD38 in PMA/ionomycin.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Quality

Evaluated by Western Blotting in Raji cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected CD38 in 10 µg of Raji cell lysate.

Target description

~35 kDa observed

Physical form

Format: Purified
Protein A purified
Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Raji cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service