Skip to Content
Merck
All Photos(2)

Key Documents

MABE326

Sigma-Aldrich

Anti-JARID1D/KDM5D Antibody, clone 7F10.4

clone 7F10.4, from mouse

Synonym(s):

JARID1D/KDM5D, Lysine-specific demethylase 5D, Histocompatibility Y antigen, H-Y, Histone demethylase JARID1D, Jumonji/ARID domain-containing protein 1D, Protein SmcY

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€395.00

€395.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€395.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€395.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

7F10.4, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KDM5D(8284)

General description

Lysine-specific demethylase 5D (UniProt Q9BY66; also known as H-Y, Histone demethylase JARID1D, Histocompatibility Y antigen, Human homolog of selected mouse cDNA on Y, Jumonji/ARID domain-containing protein 1D, Jumonji AT rich interactive domain 1D (RBP2-like), Protein SmcY, SMC homolog Y chromosome, Smcy homolog Y-linked) is encoded by the KDM5D (also known as HY, HYA, JARID1D, KIAA0234, SMCY) gene (Gene ID 8284) in human. JARID1D/KDM5D is a histone demethylase that specifically catalyzes the demethylation of tri- and di-methylated Lys4 of histone H3 (H3K4m3 and H3K4m2), but not methylated H3K9, H3K27, H3K36, H3K79, H4K20, or monomethylated H3K4. The KDM5D gene is mapped to the azoospermia factor (AZF) region known to be involved in spermatogenesis.

Specificity

Clone 7F10.4 is expected to react with all three spliced JARID1D/KDM5D isoforms.

Immunogen

KLH-conjugated linear peptide corresponding to human JARID1D/KDM5D.

Application

Immunocytochemistry Analysis: A 1:250 dilution from a representative lot detected JARID1D/KDM5D in HeLa cells.
This Anti-JARID1D Antibody, clone 7F10.4 is validated for use in Western Blotting and Immunocytochemistry for the detection of KDM5D .

Quality

Evaluated by Western Blotting in U2OS cell lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected JARID1D/KDM5D in 10 µg of U2OS cell lysate.

Target description

~200 kDa observed. Isoform 1 (174.1 kDa calculated); Isoform 2 (167.4 kDa calculated); Isoform 3 (177.6 kDa calculated). Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service