Skip to Content
Merck
All Photos(1)

Key Documents

MABC541

Sigma-Aldrich

Anti-Cullin-3 Antibody, clone 3A8.1

clone 3A8.1, from mouse

Synonym(s):

Transient receptor potential cation channel subfamily V member 4, TrpV4, Osm-9-like TRP channel 4, OTRPC4, Transient receptor potential protein 12, TRP12, Vanilloid receptor-like channel 2, Vanilloid receptor-like protein 2, VRL-2, Vanilloid receptor-rel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3A8.1, monoclonal

species reactivity

monkey, mouse

species reactivity (predicted by homology)

human (based on 100% sequence homology)

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CUL3(8452)

General description

Cullin-3 (UniProt Q13618) is encoded by the CUL3 (also known as PHA2E, KIAA0617) gene (Gene ID 8452) in human. Polyubiquitination of proteins is mediated by ubiquitin-activating enzymes (E1s), ubiquitin-conjugating enzymes (E2s), and ubiquitin ligases (E3s). There exist RING finger-type, HECT-type, and U-box type of E3 ligases. Cullin-3 belongs to the cullin family of hydrophobic proteins that confer substrate specificity to the multimeric Cullin-RING E3 ubiquitin ligase (CRL) complexes by acting as scaffold proteins. The activity of CRLs is regulated by Cullin Neddylation via the reversible conjugation of a small ubiquitin-type protein known as Neural precursor cell expressed developmentally down-regulated protein 8 (Nedd8). Cullin-3-based CRLs rely on Cullin-3 interaction with bric-a-brac/tramtrack/broad-complex (BTB) domain-containing proteins for targets recruitment.

Immunogen

Epitope: C-terminal
GST-tagged recombinant protein corresponding to the C-terminal of human Cullin-3.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-Cullin-3 Antibody, clone 3A8.1 is validated for use in Western Blotting for the detection of Cullin-3.

Quality

Evaluated by Western Blotting in mouse neuroblastoma Neuro-2a (N2a) cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Cullin-3 in 10 µg of mouse neuroblastoma Neuro-2a (N2a) cell lysate.

Target description

~88 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service