Skip to Content
Merck
All Photos(1)

Documents

MAB8920

Sigma-Aldrich

Anti-Measles Blend Antibody, hemagglutinin & matrix protein, clones CV1, CV4, CV8, CV9

ascites fluid, Chemicon®, from mouse

Synonym(s):

Rubeola

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

CV1, monoclonal
CV4, monoclonal
CV8, monoclonal
CV9, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

isotype

IgG

shipped in

wet ice

Specificity

Recognizes the measles hemagglutinin and the matrix protein by indirect immunofluorescence.

Immunogen

Epitope: hemagglutinin & matrix protein

Application

Anti-Measles Blend Antibody, hemagglutinin & matrix protein, clones CV1, CV4, CV8, CV9 detects level of Measles Blend & has been published & validated for use in IF.
Culture Confirmation.

Indirect Immunofluorescence at 1:100-1000.

Final working dilutions must be determined by end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Physical form

Ascites fluid, no preservatives.

Storage and Stability

Maintain at -20°C in aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G J Nuovo et al.
PCR methods and applications, 2(2), 117-123 (1992-11-01)
We describe a technique, called reverse transcriptase (RT) in situ PCR, whereby RNA may be nonisotopically detected in fixed cells when amplified by PCR after cDNA synthesis by RT. RT in situ PCR using primers specific for the measles virus
K G Mansfield et al.
Veterinary pathology, 51(1), 110-126 (2013-10-31)
Molecular localization techniques remain important diagnostic and research tools for the pathologist evaluating nonhuman primate tissues. In situ hybridization and immunohistochemistry protocols have been developed for many important pathogens of nonhuman primates, including RNA and DNA viruses, prions, and bacterial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service