MAB3503-I
Anti-Apaf-1 Antibody, clone 2E12
clone 2E12, from rat
Synonym(s):
Apoptotic protease-activating factor 1
About This Item
Recommended Products
biological source
rat
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
2E12, monoclonal
species reactivity
human
technique(s)
electron microscopy: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable
isotype
IgG2aκ
NCBI accession no.
UniProt accession no.
shipped in
ambient
target post-translational modification
unmodified
Gene Information
human ... APAF1(317)
General description
Specificity
Immunogen
Application
Apoptosis & Cancer
Western Blotting Analysis: A representative lot detected Apaf-1 in Western Blotting applications (Moriishi, K., et. al. (1999). Proc Natl Acad Sci USA. 96(17):9683-8).
Immunocytochemistry Analysis: A representative lot detected Apaf-1 in Immunocytochemistry applications (Hausmann, G., et. al. (2000). J Cell Biol. 149(3):623-34).
Immunoprecipitation Analysis: A representative lot detected Apaf-1 in Immunoprecipitation applications (Moriishi, K., et. al. (1999). Proc Natl Acad Sci USA. 96(17):9683-8).
Electron Microscopy Analysis: A representative lot detected Apaf-1 in Electron Microscopy applications (Hausmann, G., et. al. (2000). J Cell Biol. 149(3):623-34).
Quality
Western Blotting Analysis: 1 µg/mL of this antibody detected Apaf-1 in 10 µg of Jurkat cell lysate.
Target description
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service