Skip to Content
Merck
All Photos(1)

Documents

HTS055M

Sigma-Aldrich

ChemiSCREEN Membrane Preparation Recombinant Human CXCR5 Chemokine Receptor

Human CXCR5 GPCR membrane preparation for Radioligand binding Assays & GTPγS binding.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32161000
NACRES:
NA.84

biological source

human

Quality Level

recombinant

expressed in Chem-5 cells

manufacturer/tradename

ChemiScreen
Chemicon®

technique(s)

ligand binding assay: suitable (GTPγS)
radioligand binding assay (RLBA): suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... CXCR5(643)

General description

CXCR5 (also known as BLR1) is a GPCR that binds selectively to the chemokine CXCL13 (also known as BLC and BCA-1) to mediate immune system development and function (Förster et al., 1996; Gunn et al., 1998). CXCR5 cooperates with CCR7 to determine functional organization of lymphoid organs such as lymph nodes and Peyer’s patches (Ohl et al., 2003). In addition, CXCR5 mediates migration of B cells into the follicles of the splenic white pulp, thus permitting antigen deposition on follicular dendritic cells (Förster et al., 1996; Cinamon et al., 2008). Patients with rheumatoid arthritis (RA) displayed upregulated expression of CXCR5 to a greater extent than other chemokine receptors, and CXCR5-null mice subjected to antigen-induced arthritis display reduced joint destruction (Schmutz et al, 2004; Wengner et al, 2007). Therefore, CXCR5 represents a potential target for treatment of rheumatoid arthritis. Millipore′s CXCR5 Membrane Preparation for GTPγS Assay is a crude membrane preparation made from the proprietary stable recombinant cell line that is also used for Millipore’s CXCR5 Calcium Optimized Cell Line (HTS055C). This CXCR5 Membrane Preparation has been optimized for low basal activity in GTPγS binding assays; thus it is an ideal HTS tool for screening of compounds for agonist and antagonist activities. The membrane preparations exhibits EC50 of 1.8 nM for CXCL13 in a GTPγS binding assay.
Human CXCR5

Biochem/physiol Actions

Protein Target: CXCR5

Quality

1 unit = 5 μg
EC50 in GTPγS binding assay by CXCL13: ~ 1.8 nM

Specifications

Inucbation Conditions
Membranes are permeabilized by addition of saponin to an equal concentration by mass, then mixed with [35S]-GTPγS (final concentration of 0.3 nM) in 20 mM HEPES, pH 7.4/100 mM NaCl/10 mM MgCl2/0.5 μM GDP in a nonbinding 96-well plate. Unlabeled CXCL13 was added to the final concentration indicated in Figure 1 (final volume 100 μL), and incubated for 30 min at 30°C. The binding reaction is transferred to an FB filter plate (Millipore MAHF B1H) previously prewetted with water. The plate is washed 3 times (1 mL per well per wash) with cold 10 mM sodium phosphate, pH 7.4, then dried and counted.

One package contains enough membranes for at least 200 assays (units), where one unit is the amount of membrane that will yield greater than 1000 cpm specific CXCL13-stimulated [35S]-GTPγS binding.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service