Select a Size
€686.00
Select a Size
About This Item
€686.00
biological source
human
Quality Level
recombinant
expressed in Chem-3 cells
form
liquid
packaging
pkg of 1 mL
manufacturer/tradename
ChemiScreen
Chemicon®
technique(s)
ligand binding assay: suitable (GTPγS)
radioligand binding assay (RLBA): suitable
NCBI accession no.
UniProt accession no.
General description
Chemicon′s A3 membrane preparations are crude membrane preparations made from our proprietary stable recombinant cell lines to ensure high-level of GPCR surface expression; thus, they are ideal HTS tools for screening of antagonists of A3 interactions with its ligands. The membrane preparations exhibit a Kd of 4.7 nM for [125I]-AB-MECA. With 5 mg/well A3 Membrane Prep and 5nM [125I]-AB-MECA, a greater than 3-fold signal-to-background ratio is obtained.
Application
Biochem/physiol Actions
- GPCR Class: A
- Protein Target: A3
- Target Sub-Family: Adenosine
Quality
Specifications
Binding buffer: 50 mM Tris, pH 7.4, 10 mM MgCl2, 1 mM EDTA, filtered and stored at 4°C.
Radioligand: [125I]-AB-MECA (Perkin Elmer# :NEX312)
Wash Buffer: 50 mM Tris, pH 7.4, filtered and stored at 4°C.
One package contains enough membranes for at least 200 assays (units), where a unit is the amount of membrane that will yield greater than 3-fold signal:background with 125I-labeled AB-MECA at 0.2 nM
Physical form
Storage and Stability
Legal Information
Disclaimer
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service