Skip to Content
Merck
All Photos(2)

Key Documents

HTS046M

Sigma-Aldrich

ChemiSCREEN Membrane Preparation Recombinant Human ETB Endothelin Receptor

Human ETB GPCR membrane preparation for Radioligand binding Assays & GTPγS binding.

Sign Into View Organizational & Contract Pricing

Select a Size

200 UNITS
€686.00

€686.00


Please contact Customer Service for Availability


Select a Size

Change View
200 UNITS
€686.00

About This Item

UNSPSC Code:
41106514
eCl@ss:
32161000
NACRES:
NA.84

€686.00


Please contact Customer Service for Availability

biological source

human

Quality Level

recombinant

expressed in CHO cells

manufacturer/tradename

ChemiScreen
Chemicon®

technique(s)

ligand binding assay: suitable (GTPγS)
radioligand binding assay (RLBA): suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

General description

Human ETB isoform 1
The endothelin family of peptides are potent vasoconstrictors synthesized by endothelial cells in both constitutive and inducible pathways. The biological actions of endothelins are mediated by two GPCRs, ETA and ETB (Davenport, 2002). Whereas ETA is the predominant receptor on smooth muscle cells and thus is the primary mediator of the vasoconstrictor activity, ETB is found on endothelial cells and mediates vasodilation (Nilsson et al., 1997). In addition, ETB has also been linked to renal failure, congestive heart failure, atherosclerosis, and pulmonary hypertension (D’Orléans-Juste, et al., 2002). A mutation in ETB has been found to cause Hirschsprung disease-2, a degenerative disease of the digestive tract (Puffenberger et al, 1994). Millipore’s cloned human ETB expressing cell line is made in the CHO host, which supports high levels of recombinant ETB expression on the cell surface. Thus, the ETB Membrane Preparation is an ideal tool for screening for compounds binding to ETB. The membrane preparations exhibit a Kd of 0.03-0.06 nM for [125I]-endothelin-1. With 0.2 nM [125I]-endothelin-1, 10 μg/well ETB Membrane Prep yields greater than 5 fold signal-to-background ratio.

Application

Radioligand binding assay and GTPγS binding

Biochem/physiol Actions

GPCR Class: A
Protein Target: ETB
Target Sub-Family: Endothelin

Quality

Table 1. Signal:background and specific binding values obtained in a competition binding assay with ETB membrane prep.

10 µg/well
Signal:Background 7.4
Specific Binding (cpm) 5704


SPECIFICATIONS: 1 unit = 10 μg membrane preparation
Bmax: 0.49 pmol/mg
Kd: 0.03-0.06 nM

Specifications

Inucbation Conditions
RECOMMENDED ASSAY CONDITIONS: Membranes are mixed with radioactive ligand and unlabeled competitor (see Figures 1 and 2 for concentrations tested) in binding buffer in a nonbinding 96-well plate, and incubated for 1-2 h. Prior to filtration, an FC 96-well harvest plate (Millipore cat. # MAHF C1H) is coated with 0.33% polyethyleneimine for 30 min, then washed with 50mM HEPES, pH 7.4, 0.5% BSA. Binding reaction is transferred to the filter plate, and washed 3 times (1 mL per well per wash) with Wash Buffer. The plate is dried and counted.

Binding buffer: 50 mM Hepes, pH 7.4, 5 mM MgCl2, 1 mM CaCl2, 0.2% BSA, filtered and stored at 4°C

Radioligand: [125I]Endothelin-1 (Perkin Elmer # NEX259)

Wash Buffer: 50 mM Hepes, pH 7.4, 500mM NaCl , 0.1% BSA, filtered and stored at 4°C.

One package contains enough membranes for at least 200 assays (units), where an unit is the amount of membrane that will yield greater than 5-fold signal:background with 125I-labeled Endothelin-1 at 0.25 nM.

Physical form

One package contains enough membranes for at least 200 assays (units), where an unit is the amount of membrane that will yield greater than 5-fold signal:background with 125I-labeled Endothelin-1 at 0.25 nM.

Liquid in packaging buffer: 50 mM Tris pH 7.4, 10% glycerol and 1% BSA with no preservatives.
Packaging method: Membrane protein was adjusted to the indicated concentration in packaging buffer, rapidly frozen, and stored at -80°C.

Storage and Stability

Store at –70°C. Product is stable for at least 6 months from the date of receipt when stored as directed. Do not freeze and thaw.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service