Skip to Content
Merck
All Photos(1)

Key Documents

HCKP1-11K-PXBK17

Millipore

MILLIPLEX® Human Immuno-Oncology Checkpoint Protein Premixed 17-plex Panel 1 Space Saver (Bulk) Packaging - Immuno-Oncology Multiplex Assay

Synonym(s):

17-Plex Immuno-Oncology Kit, Human Checkpoint Protein Panel, Immuno-Oncology 17-Plex Panel

Sign Into View Organizational & Contract Pricing

Select a Size

HCKP1-11K-PXBK17
€3,330.00

€3,330.00


Product Availability: You will receive an order acknowledgement once the order has been entered in our system.

Request more information

Select a Size

Change View
HCKP1-11K-PXBK17
€3,330.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

€3,330.00


Product Availability: You will receive an order acknowledgement once the order has been entered in our system.

Request more information

species reactivity

human

manufacturer/tradename

Milliplex®

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex® xMAP®)

storage temp.

2-8°C

General description

Immune checkpoint proteins are regulators of immune activation and play a key role in maintaining immune homeostasis, preventing autoimmunity, and are involved in cancer immune escape. These immune system molecules demonstrate the ability to act both as agonists (co-stimulatory checkpoint molecules) or antagonists (co-inhibitory checkpoint molecules) of immune response. Immune checkpoint inhibitor drugs targeting inhibitory immune checkpoint proteins, such as CTLA-4 and PD-L1, have demonstrated efficacy in cancer immunotherapy by improving the antitumor immune response and saving lives in the treatment of many malignancies. Immune checkpoint proteins may express on immune cells and/or tumor cells as activating receptors or inhibitory receptors. Many immune checkpoint proteins are also expressed as soluble forms in circulation and in the tumor and tumor microenvironment and are putative biomarkers for immunotherapy in various diseases including cancer.The MILLIPLEX® Human Immuno-Oncology Checkpoint Protein Panel 1 is a 17-plex kit to be used for the simultaneous quantification of all of the following analytes in serum, plasma and tissue culture samples: BTLA (B- and T-lymphocyte attenuator), CD27 (Cluster of differentiation 27), CD28 (Cluster of differentiation 28), CD40 (Cluster of differentiation 40), CD80/B7-1 (Cluster of differentiation 80/B7-1), CD86/B7-2 (Cluster of differentiation 86/B7-2), CTLA-4 (cytotoxic T-lymphocyte-associated protein 4), GITRL (Ligand for receptor TNFRSF18/AITR/GITR), GITR (Glucocorticoid-induced TNFR-related protein), HVEM (Herpesvirus entry mediator), ICOS (Inducible T-cell costimulator), LAG-3 (Lymphocyte-activation gene 3), PD-L1 (Programmed death-ligand 1), PD-L2 (Programmed death-ligand 2), PD-1 (Programmed cell death protein 1), TIM-3 (T-cell immunoglobulin and mucin-domain containing-3), TLR-2 (Toll like receptor 2).

Specificity

Cross-reactivity between the antibodies and any of the other analytes in this panel is non-detectable or negligible.

Application

The Human Immuno-Oncology Checkpoint Protein Panel 1 bead-based multiplex panel, using the Luminex® xMAP technology, enables the simultaneous analysis of 17 immune checkpoint proteins in human serum, plasma and cell culture samples.For optimal sensitivity, use an overnight (16-18 hour) incubation. Serum and plasma samples require (recommended) a 1:2 dilution with Assay Buffer. Tissue culture supernatants may be run neat or appropriately diluted in control medium. Each well requires 25 μL of sample, standard or control.Analytes available:BTLA;CD27;CD28;CD40;CD80/B7-1;CD86/B7-2;CTLA-4;GITR;GITRL;HVEM;ICOS;LAG-3;PD-1;PD-L1;PD-L2;TIM-3;TLR-2

Features and Benefits

Configuration: Premixed

Packaging

96-well plate

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service