HAEP04700
MCE Membrane Filter, 0.45 μm Pore Size
MF-Millipore™, filter diam. 47 mm, hydrophobic
Synonym(s):
MF-Millipore™ Membrane Filter, Hydrophobic Edge, 0.45 µm pore size
About This Item
Recommended Products
material
mixed cellulose esters (MCE) membrane
plain filter
white filter
Quality Level
description
47 mm diameter, mixed cellulose esters (MCE) membrane w/ 3 mm hydrophobic edge, white, 100 discs
feature
hydrophobic
manufacturer/tradename
MF-Millipore™
Millipore
parameter
4 L/min-cm2 air flow rate
60 mL/min-cm2 water flow rate
75 °C max. temp.
filter diam.
47 mm
thickness
150 μm
color
white
refractive index
n/D 1.51
matrix
MF-Millipore™
pore size
0.45 μm pore size
79 % porosity
bubble point
≥1.8 bar, air with water at 23 °C
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
Application
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Sol. 1
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service