Skip to Content
Merck
All Photos(1)

Documents

GSWP047S0

Millipore

SO-Pak Filters 0.22µm 47mm white plain

Microbiological analysis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24

material

mixed cellulose esters (MCE)
plain filter
white filter

sterility

sterile; ethylene oxide treated

feature

hydrophilic

packaging

box of 10 envelopes containing 10 pak

filter diam.

47 mm

pore size

0.22 μm pore size

input

sample type bottled water
sample(s) (environmental)
sample type beer
sample type cosmetics
sample type beverage(s)
sample type food(s)
sample type soft drink
sample type cider
sample type pharmaceutical(s)
sample type wine
sample type sport drink

application(s)

bioburden testing
clinical
cosmetics
environmental
food and beverages
industrial qc
pharmaceutical

shipped in

ambient

General description

SO Pak filters - 10 resealable envelopes containing 10 filters and 10 pads, presterilized.

Application

Microbiological analysis

Packaging

Sterile pack: 10 resealable envelopes containing 10 filters and 10 pads, sterile

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service