Skip to Content
Merck
All Photos(1)

Key Documents

GF426

Sigma-Aldrich

IL-28B, Human

HumanKine®, recombinant, Xeno-free, >95% (SDS-PAGE), suitable for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.75

product name

HumanKine® IL-28B, Human Recombinant Xeno-Free,

Assay

>95% (SDS-PAGE)

Quality Level

feature

endotoxin free

UniProt accession no.

General description

HumanKine growth factors are expressed in a proprietary human 293 protein expression system and are manufactured without the use of any animal products and are considered Xeno-Free.
IL-28B is a cytokine distantly related to type I interferons and the IL-10 family. IL-28A, IL-28B, and IL-29 are three closely related cytokine genes that form a cytokine gene cluster on a chromosomal region mapped to 19q13. Expression of the cytokines encoded by the three genes can be induced by viral infection. All three cytokines have been shown to interact with a heterodimeric class II cytokine receptor that consists of interleukin 10 receptor, beta (IL10RB) and interleukin 28 receptor, alpha (IL28RA).

Quality

Bioactivity assay: Determined by the dose dependent protection of the cytopathic effect on A549 cells (human lung
adenocarcinoma epithelial cell line) that were challenged with encephalomyocarditis (EMC) virus. ED50 is typically 0.1-1 ng/ml.
HumanKine IL-28B is manufactured using all non-animal reagents and is considered Xeno-Free

Physical form

Product is presented in PBS and filtered through a 0.2 micron filter before lyophilization.

Storage and Stability

Store at -20°C for up to 4 months from date of receipt. Further dilutions should be made into appropriate buffer or medium to which protein has been added.

Legal Information

HumanKine is a registered trademark of Proteintech Group, Inc. and Humanzyme, Inc

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service