Skip to Content
Merck
All Photos(1)

Documents

GF128

Sigma-Aldrich

Stromal Cell-Derived Factor-1α Protein, Recombinant mouse

Stromal Cell-Derived Factor-1 alpha protein (or SDF-1 alpha protein) is a recently discovered protein belonging to the alpha chemokine (CXC) family of cytokines.

Synonym(s):

SDF-1alpha

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.77

biological source

mouse

Quality Level

Assay

>99% (SDS-PAGE and HPLC)

manufacturer/tradename

Chemicon®

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 ng/μg Endotoxin (of SDF-1 alpha; 1EU/ug)

input

sample type hematopoietic stem cell(s)

UniProt accession no.

shipped in

dry ice

General description

Product Source: E.coli
Stromal Cell-Derived Factor-1 alpha (SDF-1 alpha) is a recently discovered protein belonging to the alpha chemokine (CXC) family of cytokines. Recombinant Mouse SDF-1 alpha is a 7.9 kDa protein containing 68 amino acid residues.

Physical form

The sterile filtered solution was lyophilized with no additives.

Storage and Stability

The lyophilized powder is stable for a few weeks at room temperature, but best stored at -20°C. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. This solution can then be diluted into other aqueous buffers and stored at 4°C for 1 week or -20°C for future use. Reconstituted mouse SDF-1 alpha should be stored in undiluted aliquots at -20°C. Avoid repeated freeze/thaw cycles.

Analysis Note

Specific Activity: Determined by its ability to chemoattract human peripheral blood monocytes using a concentration of 50-100ng/ml.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service