Skip to Content
Merck
All Photos(1)

Key Documents

GC0004

Supelco

Trifluoroacetic acid solution

10%

Synonym(s):

Trifluoroacetic acid solution, acid, TFA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3COOH
CAS Number:
Molecular Weight:
114.02
Beilstein:
5446934
MDL number:
UNSPSC Code:
12191502
Product GC0004 is not currently sold in your country. Contact Technical Service

Product Name

Trifluoroacetic acid solution, 0.1%

Quality Level

form

liquid

shelf life

18 mo. (from manufacture date)

composition

Trifluoroacetic acid, 0.095-0.105 % (v/v)

color

APHA: ≤10

shipped in

ambient

storage temp.

no temp limit

SMILES string

OC(=O)C(F)(F)F

InChI

1S/C2HF3O2/c3-2(4,5)1(6)7/h(H,6,7)

InChI key

DTQVDTLACAAQTR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Expiration date18 months from mfg date% V/ V Trifluoroacetic acid0.095-0.105 % V/VColor (APHA)10APHA maxFormClear liquidpH at 25°CTo be recorded

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service