Skip to Content
Merck
All Photos(1)

Key Documents

FCMAB174F

Sigma-Aldrich

Milli-Mark® Anti-CD7-FITC Antibody, clone CBC.37

clone CBC.37, Milli-Mark®, from mouse

Synonym(s):

T-cell antigen CD7, GP40, T-cell leukemia antigen, TP41, T-cell surface antigen Leu-9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.44

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified antibody

antibody product type

primary antibodies

clone

CBC.37, monoclonal

species reactivity

human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CD7(924)

General description

The CD7 antigen is a membrane-bound 40 kDa glycoprotein expressed on thymocytes, mature T cells, a large majority of natural killer cells, pluripotent hematopoietic stem cells, and progenitor cells of lymphoid and myeloid cells (Bowen, 1997 & 2002; Reiter, 1989; Chang, 1994). CD7 is one of the earliest T-cell-specific antigens to be expressed in lymphocytes, and the only early marker to persist throughout differentiation (Bowen, 1997).

Application

Detect CD7 using this Milli-Mark Anti-CD7-FITC Antibody, clone CBC.37 validated for use in FC.

Quality

Evaluated by flow cytometry using human PBMCs

Target description

40 kDa (Bowen, 2002; Reiter, 1989; Chang, 1994)

Physical form

Purified Mouse monoclonal IgG2b conjugated to FITC in 0.05M Tris-HCl buffer with 1% BSA, Sodium Azide at 0.097% at pH 7.2.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service