Skip to Content
Merck
All Photos(1)

Key Documents

EZHPP40K

Millipore

Human Pancreatic Polypeptide (PP) ELISA

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€913.00

€913.00


Estimated to ship onMay 14, 2025Details


Request a Bulk Order

Select a Size

Change View
1 PKG
€913.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.77

€913.00


Estimated to ship onMay 14, 2025Details


Request a Bulk Order

Quality Level

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service