Skip to Content
Merck
All Photos(1)

Key Documents

EM0182

Supelco

Buffer pH 1.68

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161703

grade

analytical standard

Quality Level

Agency

traceable to NIST

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

format

mixture

shipped in

ambient

storage temp.

15-25°C

Application

This pH 1.68 buffer solution is appropriate for use in routine analytical procedures for the calibration of pH meters and maintaining the pH of solutions.

Features and Benefits

  • Traceable to NIST standards
  • A ready-to-use solution available in a sealed bottle with convenient packaging to protect against contamination
  • Accompanied with a certificate of analysis (CoA) for an effective audit
  • Accurate up to +/- 0.01 at 25 °C
  • The expiry date of the product can be found in the CoA
  • Calculated uncertainty value is for the duration of transportation and storage

Specifications

Tolerance: ±0.01

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service