Skip to Content
Merck
All Photos(1)

Documents

ECM556

Sigma-Aldrich

QCM Collagen Cell Invasion Assay, 96-well (8 µm), fluorimetric

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.32

species reactivity

vertebrates

Quality Level

technique(s)

cell based assay: suitable

detection method

fluorometric

Application

The CHEMICON Cell Invasion Assay Kit is ideal for evaluation of invasive tumor cells. Each CHEMICON Cell Invasion Assay Kit contains sufficient reagents for the evaluation of 96 samples. The quantitative nature of this assay is especially useful for large scale screening of pharmacological agents.

The CHEMICON Cell Invasion Assay Kit is intended for research use only; not for diagnostic or therapeutic applications.

Packaging

96 wells

Components

Sterile 96-well Cell Invasion Plate Assembly: (Part No. 90249) One 96-well feeder tray, and one 96-well Cell Invasion Chamber plate with collagen-coated inserts. 96-well Cell Culture Tray: (Part No. 90129) One 96-well feeder tray. Cell Detachment Solution: (Part No. 90131) One bottle - 16 mL. 4X Cell Lysis Buffer: (Part No. 90130) One bottle - 16 mL. CyQuant GR Dye1: (Part No. 90132) One vial - 75 mL

Storage and Stability

Store kit materials at 2-8°C for up to their expiration date. Do not freeze.

Legal Information

GR is a trademark of Sigma-Aldrich Co. LLC

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service