Skip to Content
Merck
All Photos(1)

Key Documents

E8035-HS

Sigma-Aldrich

GLP-1 HS ELISA Standard

Sign Into View Organizational & Contract Pricing

Select a Size

E8035-HS
€248.00

€248.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
E8035-HS
€248.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.84

€248.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service