The solubility of DX0145, Dextrose, is as follows: it is soluble in water (0.9 g/ml at 25°C), alcohol (slightly), methanol (at 20°C), absolute alcohol (very sparingly), ether (very sparingly), acetone (very sparingly), hot glacial acetic acid, pyridine, aniline, ethanol (<1 mg/mL at 25°C), and DMSO (36 mg/mL at 25°C).
DX0145-5
Dextrose
Synonym(s):
D-(+)-Glucose, D-Glucose, Corn sugar, Dextrose
About This Item
Recommended Products
form
solid
color
white
mp
150-152 °C (lit.)
solubility
470 g/L
shipped in
ambient
SMILES string
OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O
InChI
1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6+/m1/s1
InChI key
WQZGKKKJIJFFOK-DVKNGEFBSA-N
Looking for similar products? Visit Product Comparison Guide
General description
DX0145-3 2.5kg Poly Bottle
Application
Analysis Note
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
What is the solubility of DX0145, Dextrose?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service