Key Documents
CDRCHKIT2
Milli-Q
Reverse Osmosis Cartridge
High fouling RO Kit for RiOs 50 and Elix® 35 Water Purification System
Select a Size
1 EA
inquire
This item is not able to be purchased as a guest user. For further information, please contact us at 916572072.
Select a Size
Change View
1 EA
inquire
About This Item
UNSPSC Code:
41104214
NACRES:
JA.13
This item is not able to be purchased as a guest user. For further information, please contact us at 916572072.
packaging
pkg of 1 unit
compatibility
for use with Elix®
for use with RiOs
General description
The Reverse Osmosis membrane removes 95%-99% of contaminants including ions, particulates, bacteria, and organic molecules. For use with, Elix® 35 and RiOs 50 water purification systems.
Application
The RO water delivered from Elix® and RiOs systems is used in laboratories, mainly for applications such as general rinsing, feedwater to washing machines and autoclaves, or as a feed to pure and ultrapure water purification systems.
Legal Information
ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany
RIOS is a trademark of Merck KGaA, Darmstadt, Germany