Skip to Content
Merck
All Photos(1)

Key Documents

CC1048

Sigma-Aldrich

MMP-9, human, proform

Synonym(s):

Progelatinase B, 92 kDa Type IV Collagenase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
eCl@ss:
32160405
NACRES:
NA.41

biological source

human

Assay

>95% (total protein)

form

liquid

specific activity

>1200 mU/mg (specific activity of activated MMP-9 after trypsin activation)

manufacturer/tradename

Chemicon®

concentration

0.2 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... MMP9(4318)

General description

When measured with the peptide substrate dinitrophenyl-Pro-Gln-Gly-Ile-Ala-Gly-Gln-D-Arg [Masui et al., 1977] the specific activity of MMP-9 monomer

Application

ACTIVATION:

An aliquot of 10 μL MMP-9 monomer is mixed with 20 μL trypsin solution (see below) and activation buffer in a total volume of 100 μL. The mixture is incubated for 20 min at 37°C. Thereafter trypsin is inhibited by addition of 10 μL aprotinin solution.

Trypsin solution: 0.50 mg TPCK-trypsin/mL activation buffer. The solution is stored in aliquots at -20°C.

Activation Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 5 mM CaCl2.

Aprotinin solution: 1 mg aprotinin/mL activation buffer. The solution is stored at -20°C.

Unit Definition

Specific Activity: where 1 U is the activity that hydrolyzes 1 mmol peptide (7-methoxycoumarin-4-yl)acetyl-Pro-Leu-Gly-Leu-Dpa-Ala-Arg) within 1 minute under the assay conditions described by Knight ,et al

Physical form

Provided as a liquid in 50 mM Tris-HCl, pH 7.0, 200 mM NaCl, 5 mM CaCl2, ,1 μM ZnCl2, 0.05% Brij-35, 0.05% NaN3.

Storage and Stability

Maintain frozen at -70°C in undiluted aliquots. The enzyme may be stored at -20°C for several without significant loss of activity. Repeated freezing and thawing should be avoided.

Analysis Note

MMP-9 monomer appears as a major band at 92 kDa in non-reducing SDS-PAGE

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Secretion of collagenolytic enzymes by human polymorphonuclear leukocytes.
Hibbs, M S, et al.
Collagen and Related Research, 4, 467-477 (1984)
Matrix metalloproteinases (MMP-2 and MMP-9) of the oral cavity: cellular origin and relationship to periodontal status.
Makela, M, et al.
Journal of Dental Research, 73, 1397-1406 (1994)
I E Collier et al.
Genomics, 9(3), 429-434 (1991-03-01)
The 72- and 92-kDa type IV collagenases are members of a group of secreted zinc metalloproteases. Two members of this family, collagenase and stromelysin, have previously been localized to the long arm of chromosome 11. Here we assign both of
M(r) 92,000 type IV collagenase is increased in plasma of patients with colon cancer and breast cancer.
Zucker, S, et al.
Cancer Research, 53, 140-146 (1993)
Gelatinases A and B.
Murphy, G and Crabbe, T
Test, 248, 470-484 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service