Skip to Content
Merck
All Photos(3)

Key Documents

CBL270-I

Sigma-Aldrich

Anti-TUBA1A (TUBA3) Antibody

rat monoclonal, YOL1/34

Synonym(s):

Tubulin alpha-1 chain, alpha-tubulin

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€405.00

€405.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€405.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€405.00


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

Anti-alpha-Tubulin Antibody, clone YOL1/34, clone YOL1/34, from rat

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

YOL1/34, monoclonal

species reactivity

rat, mouse, human, yeast, amoeba, porcine

technique(s)

ELISA: suitable
affinity binding assay: suitable
electron microscopy: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

Saccharomyces cerevisiae ... Tub1(854889)
human ... TUBA1A(7846)

General description

Tubulin alpha-1 chain (UniProt P09733; also known as alpha-tubulin) is encoded by the TUB1 gene (Gene ID 854889) in yeast. Microtubules are dynamic protein filaments involved in a wide variety of cellular activities ranging from mitosis and transport events to cell movement and the maintenance of cell shape. Microtubules are assembled from α- and β-tubulin subunits, both of which are subjected to various post-translational modifications (PTMs), including acetylation, polyglutamylation, polyglycylation, detyrosination, phosphorylation, and palmitoylation. These PTMs can regulate the polymerization properties of tubulins and/or their interactions with microtubule associated proteins (MAPs) and motor proteins. Because the C-terminus tyrosine/phenylalanine is preceded by glutamate (E) residues, the detyrosinated/dephenylalaninated α-tubulin is also referred to as Glu-tubulin. While tyrosine can be reattached to the C-terminus of the Glu-tubulin by tubulin-tyrosine ligase in mammals, phenylalanine is not added posttranslationally to the Glu-tubulin in yeast despite the presence of the yeast YBR094w gene that exhibits significant homology to tubulin-tyrosine ligase in other organisms

Specificity

Clone YOL1/34 reacts with alpha-tubulin, but not beta-tubulin, by targeting a C-terminal epitope highly conserved among alpha-tubulin subtypes. Epitope is not present in alpha-tubulin 4B (TUBA4B; UniProt Q9H853).
Expected to react with a broad range of species based on 100% sequence homology.

Immunogen

Yeast tubulin (Kilmartin, J.V., et al. (1982). J. Cell Biol. 93(3):576-582).

Application

Immunocytochemistry Analysis: A 1:200 dilution from a representative lot detected alpha-tubulin in HeLa cell lysate.

Western Blotting Analysis: 1 µg/mL from a representative lot detected alpha-tubulin in 10 µg of NIH/3T3 mouse fibroblast and PC12 rat pheochromocytoma cell lysates.

Affinity Binding Assay: Pig brain α-tubulin, α-tubulin/β-tubulin heterodimer, as well as α-tubulin proteolytic fragments a.a. 414-425 and a.a. 403-422, but not β-tubulin or actin, effectively competed against immobilized α-tubulin for YOL 1/34 binding (Breitling, F., and Little, M. (1986). J. Biol. Chem. 189(2):367-370).

Electron Microscopy Analysis: A representative lot immunostained microtubules of crude yeast nuclear envelope preparation (Kilmartin, J.V., et al. (1982). J. Cell Biol. 93(3):576-582).

ELISA Analysis: A representative lot detected pig brain α-tubulin by direct (non-sandwich) ELISA (Breitling, F., and Little, M. (1986). J. Biol. Chem. 189(2):367-370).

Immunocytochemistry Analysis: A representative lot immunostained fixed NIH/3T3 cells as well as yeast spheroplasts and nuclei preparations (Kilmartin, J.V., et al. (1982). J. Cell Biol. 93(3):576-582).

Immunocytochemistry Analysis: A representative lot immunostained microtubules of fixed Dictyostelium (amoeba) as well as isolated and stabilized amoeba cytoskeleton (Kilmartin, J.V., et al. (1982). J. Cell Biol. 93(3):576-582).

Western Blotting Analysis: A representative lot detected α-tubulin in spermatozoa sample from from fertile and infertile men (Breitling, F., et al. (1991). Gene. 104(2):147-153).
This rat monoclonal Anti-alpha-Tubulin Antibody, clone YOL1/34, Cat. No. CBL270-I is validated for use in Affinity Binding Assay, Electron Microscopy, ELISA, Immunocytochemistry, and Western Blotting for the detection of alpha-tubulin.

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected alpha-tubulin in 10 µg of HeLa cell lysate.

Target description

~50 kDa observed. 50.14/46.30 kDa (human alpha-1A isoform 1/2; TUBA1A; TUBA3), 50.15/37.22 kDa (human alpha-1B isoform 1/2; TUBA1B), 44.96 kDa (human alpha-2; TUBA2; TUBA3C; TUBA3D), 49.92/48.33 kDa (human alpha-4A isoform 1/2; TUBA4A; TUBA1), 50.14 kDa (mouse & rat alpha-1A; Tuba1a; Tuba1), 50.15 kDa (mouse & rat alpha-1B; Tuba1b; Tuba2), 49.96 kDa (mouse & rat alpha-3; Tuba3; Tuba3a; Tuba3b), 49.92 kDa (mouse & rat alpha-4A; Tuba4a; Tuba4), 49.80 kDa (yeast) calculated. Uncharacterized bands may be observed in some lysate(s).

Linkage

Replaces: CBL270

Physical form

Format: Purified
Purified rat IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shota Hiruma et al.
FEBS letters, 591(20), 3296-3309 (2017-09-11)
The molecular mechanism that governs cytoskeleton-membrane interaction during animal cytokinesis remains elusive. Here, we investigated the dynamics and functions of ERM (Ezrin/Radixin/Moesin) proteins during cytokinesis in human cultured cells. We found that ezrin is recruited to the cleavage furrow through
Marc Duque et al.
Nature communications, 13(1), 600-600 (2022-02-11)
Ultrasound has been used to non-invasively manipulate neuronal functions in humans and other animals. However, this approach is limited as it has been challenging to target specific cells within the brain or body. Here, we identify human Transient Receptor Potential
Félix Matadamas-Martínez et al.
Memorias do Instituto Oswaldo Cruz, 115, e190348-e190348 (2020-02-13)
It was previously demonstrated that CMC-20, a nitazoxanide and N-methyl-1H-benzimidazole hybrid molecule, had higher in vitro activity against Giardia intestinalis WB strain than metronidazole and albendazole and similar to nitazoxanide. To evaluate the in vitro activity of CMC-20 against G.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service