Skip to Content
Merck
All Photos(1)

Key Documents

ATBFEET01

Millipore

M Air T Isolator Tripod

suitable for air monitoring, M Air T

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€963.00

€963.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 EA
€963.00

About This Item

UNSPSC Code:
41104011
eCl@ss:
25140604
NACRES:
NB.11

€963.00


Please contact Customer Service for Availability

Request more information

material

stainless steel body

manufacturer/tradename

M Air T

input

sample type pharmaceutical(s)

application(s)

air monitoring
environmental
pharmaceutical

compatibility

for use with M Air T Compressed Gas and Air Test Kit
for use with M Air T Isolator

shipped in

ambient

Legal Information

M AIR T is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service