Skip to Content
Merck
All Photos(1)

Key Documents

AG952

Sigma-Aldrich

Tau-381, recombinant human

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€622.00

€622.00


Please contact Customer Service for Availability


Select a Size

Change View
50 μG
€622.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.42

€622.00


Please contact Customer Service for Availability

biological source

human

Quality Level

Assay

>90% (SDS-PAGE)

form

powder

manufacturer/tradename

Chemicon®

UniProt accession no.

shipped in

dry ice

General description

Tau-381

MOL. WT.: 39,700 Da

PEPTIDE CONTENT: 50 μg per vial (protein content and concentration determined by BCA protein assays using BSA as standard).

COUNTER ION: Lyophilized in final buffer of 50 mM MES, pH 6.8, 100 mM NaCl and 0.5 mM EGTA.

Application

Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Physical form

White lyophilized powder. Resuspend in water at a concentration of 1 mg/mL. This will give you a final of 50 mM MES, pH 6.8, 100 mM NaCl and 0.5 mM EGTA.

Storage and Stability

Maintain lyophilized material at -20ºC for up to 12 months after date of receipt. After reconstitution maintain at -20ºC to -70ºC for up to 2 weeks in undiluted aliquots. Avoid freeze/thaw cycles to avoid aggregation.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohamed B Abou-Donia et al.
Brain sciences, 10(9) (2020-09-10)
For the past 30 years, there has been a lack of objective tools for diagnosing Gulf War Illness (GWI), which is largely characterized by central nervous system (CNS) symptoms emerging from 1991 Gulf War (GW) veterans. In a recent preliminary
Mohamed B Abou-Donia et al.
Brain sciences, 11(2) (2021-01-28)
Veterans from the 1991 Gulf War (GW) have suffered from Gulf War illness (GWI) for nearly 30 years. This illness encompasses multiple body systems, including the central nervous system (CNS). Diagnosis and treatment of GWI is difficult because there has

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service