Skip to Content
Merck
All Photos(1)

Key Documents

AG334

Sigma-Aldrich

Opioid Receptor, δ 358-372, control peptide for AB5505

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€186.00

€186.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
20 μG
€186.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.25

€186.00


Please contact Customer Service for Availability

Request a Bulk Order

technique(s)

cell based assay: suitable

Quality Level

UniProt accession no.

Gene Information

General description

FUNCTION: SwissProt: P41143 # Inhibits neurotransmitter release by reducing calcium ion currents and increasing potassium ion conductance. Highly stereoselective. receptor for enkephalins.
SIZE: 372 amino acids; 40413 Da
SUBUNIT: Interacts with GPRASP1.
SUBCELLULAR LOCATION: Cell membrane; Multi-pass membrane protein.
SIMILARITY: SwissProt: P41143 ## Belongs to the G-protein coupled receptor 1 family.

Application

For use in blocking the reactivity of AB5505.

Optimal working concentration must be determined by the end user.

Physical form

Lyophilized from PBS, pH 7.4. Reconstitute with 20 μL of sterile distilled water.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles. During shipment, small volumes of product will occasionally become entrapped in the seal of the product vial. For products with volumes of 200 μL or less, we recommend gently tapping the vial on a hard surface or briefly centrifuging the vial in a tabletop centrifuge to dislodge any liquid in the container′s cap.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service