Skip to Content
Merck
All Photos(1)

Key Documents

AG217

Sigma-Aldrich

Histamine Receptor 4, control peptide for AB5663P

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€429.00

€429.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€429.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.25

€429.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

human

Quality Level

form

liquid

manufacturer/tradename

Chemicon®

concentration

1.0 mg/mL

technique(s)

cell based assay: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

General description

Synthetic 17 amino acid peptide sequence within the cytoplasmic, C-terminus of human HR4.

Application

For use in blocking the reactivity of AB5663P.

Optimal working dilution must be determined by the end user.
Research Category
Signaling
Research Sub Category
Neurotransmitters & Receptors

Physical form

Liquid in PBS containing 0.1% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service