Skip to Content
Merck
All Photos(1)

Key Documents

ABT253

Sigma-Aldrich

Anti-Periostin Antibody (C-term)

1 mg/mL, from rabbit

Synonym(s):

Periostin, PN, Osteoblast-specific factor 2, OSF-2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€347.00

€347.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€347.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€347.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

concentration

1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... POSTN(10631)

General description

Periostin, also known as PN, or Osteoblast-specific factor 2 (OSF-2), and encoded by the gene POSTN/OSF2, is a secreted extracellular matrix protein that appears to be involved in inducing cell attachment and spreading as well as cell adhesion. In bone, Periostin enhances mineralization and assists in the incorporation of BMP1 protein into the fibronectin extracellular matrix of connective tissues. Periostin is preferentially expressed in periosteum and periodontal ligaments but can also be found expressed in developing heart and other tissues. Periostin is also over expressed in many cancers where is contributes to a growth-supportive role for such cancers and also aids in chemoresistance for cancers cells through the activation of the PI3K/Akt/surviving pathway.

Specificity

This antibody detects a subset of splice variants of Periostin.

Immunogen

A 188-amino acid bacterial fusion protein from the C-terminal region of mouse periostin.
Epitope: C-terminus

Application

Anti-Periostin Antibody (C-term) is an antibody against Periostin (C-term) for use in western blotting.
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)

Quality

Evaluated by Western Blotting in mouse lung tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Periostin in 10 µg of mouse lung tissue lysate.

Target description

~80-85 kDa observed. Multiple splice variants detected.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service