ABT1385M
Anti-Glypican-1
serum, from rabbit
Synonym(s):
HSPG M12, GPC1
Select a Size
Select a Size
About This Item
Recommended Products
biological source
rabbit
antibody form
serum
antibody product type
primary antibodies
clone
polyclonal
species reactivity
rat
packaging
antibody small pack of 25 μL
technique(s)
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable
isotype
IgG
NCBI accession no.
UniProt accession no.
target post-translational modification
unmodified
Gene Information
rat ... Gpc1(58920)
General description
Specificity
Immunogen
Application
Cell Structure
Immunocytochemistry Analysis: A representative lot detected Glypican-1 in rat C6 glioma cells (Liang, Y., et. al. (1997). J Cell Biol. 139(4):851-64).
Immunohistochemistry Analysis: A representative lot detected Glypican-1 in rat spinal cord and brain (Liang, Y., et. al. (1997). J Cell Biol. 139(4):851-64).
Quality
Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected Glypican-1 in Rat C6 glioma cells.
Target description
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service