Skip to Content
Merck
All Photos(2)

Key Documents

ABS311

Sigma-Aldrich

Anti-LCN13 Antibody

serum, from rabbit

Synonym(s):

orant-binding protein 2a, Epididymal-specific lipocalin-13

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€326.00

€326.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
€326.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€326.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Obp2A(227627)

General description

Epididymal-specific lipocalin-13 (LCN13) is also called Odorant-binding protein 2a (Obp2a). LCN13 is a secreted protein that is thought to act as a retinoid carrier protein within the epididymis, and have a role in male fertility. LCN13 is expressed in the epididymis.

Specificity

This antibody detects two observed isoforms of LCN13.

Immunogen

Recombinant protein corresponding to mouse LCN13.

Application

Detect Lipocalin using this rabbit polyclonal antibody, Anti-LCN13 Antibody validated for use in western blotting.
Research Category
Metabolism
Research Sub Category
Lipid Metabolism & Weight Regulation
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected LCN13 in mouse lung tissue lysate.

Western Blotting Analysis: A representative lot from an independent laboratory detected LCN13 in liver, muscle, and pancreas tissues, but not in white adipose tissue, from LCN13 transgenic male mice (Cho, K. W., et al. (2011). Mol Cell Biol. 31(3):450-7.).

Western Blotting Analysis: A representative lot from an independent laboratory detected LCN13 in transfected mouse primary hepotcytes cell lysates (Sheng, L, et al. (2011). J Biol Chem. 286(44):38128-38135.).

Western Blotting Analysis: A representative lot detected LCN13 in mouse adipocyte cell lysates and in certain mouse organ tissues lysates (Zhou, Y. and Rui, L., (2013). Vitam Horm. 91:369-383.).

Quality

Evaluated by Western Blotting in mouse skin tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected LCN13 in 10 µg of LCN13 mouse skin tissue lysate.

Target description

~20 kDa observed. Two observed isoforms at ~20 kDa and ~28 kDa are likely due to proteolytic cleavage of a common LCN13 precursor (Cho, K. W., et al. (2011). Mol Cell Biol. 31(3):450-7.). Uncharacterized band(s) may be observed in some cell lysates.

Physical form

Rabbit polyclonal serum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Mouse skin tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service