Skip to Content
Merck
All Photos(1)

Key Documents

ABS1030

Sigma-Aldrich

Anti-Apolipoprotein D Antibody

serum, from rabbit

Synonym(s):

ApoD, Apolipoprotein D, Apo-D

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€323.00

€323.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
€323.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€323.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... APOD(347)

General description

Apolipoprotein D (ADOP), also known as Apolipoprotein D, Apo-D, ApoD, and encoded by the gene name APOD, is a member of the calycin superfamily and the Lipocalin family. Recent studies have suggested that Apolipoprotein D is involved in the transport and binding of bilin. Additioanlly, studies have demonstrated that Apolipoprotein D is able to transport a variety of ligands in a number of different contexts. Apolipoprotein D is a homodimer located in the plasma, and it also exists as a disulfide-linked heterodimer with APOA2. Apolipoprotein D is expressed in the liver, intestine, pancreas, kidney, placenta, adrenal, spleen, fetal brain tissue and tears. Apolipoprotein D is primarily localized in HDL (60-65%), with most of the remainder in VHDL and only small amounts in VLDL and LDL.

Immunogen

Protein purified from human plasma corresponding to Apolipoprotein D.

Application

Anti-Apolipoprotein D is an antibody against Apolipoprotein D for use in ELISA, WB.
ELISA Analysis: A representative lot of this antibody detected endogenous Apolipoprotein D in normal human plasma (Knipping, G., et al. Journal of Immunological Methods 202 1997. 85–95)
Western Blotting Analysis: A representative lot of this antibody at 1:5000 dilution detected endogenouse Apolipoprotein D in 5 µl of human plasma (Bottcher, A., et al. J. Lipid Res. 2000. 41: 905–915.)
Research Category
Signaling
Research Sub Category
Signaling Neuroscience

Quality

Evaluated by Western Blotting on purified human Apolipoprotein D

Western Blotting Analysis: A 1:1000 dilution of this antibody detected 0.05 µg of purified human Apolipoprotein D

Target description

~30 kDa observed, additional bands likely due to heavy N-glycosylation on this protein

Physical form

Rabbit polyclonal serum with 0.05% sodium azide.
Unpurified

Storage and Stability

"Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance."

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service