Skip to Content
Merck
All Photos(1)

Key Documents

ABN2252

Sigma-Aldrich

Anti-Neuroligin-4

Synonym(s):

Neuroligin Y

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€138.00

€138.00


Estimated to ship onMarch 20, 2025Details


Request a Bulk Order

Select a Size

Change View
25 μG
€138.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€138.00


Estimated to ship onMarch 20, 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

species reactivity

human

packaging

antibody small pack of 100

technique(s)

immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal cytoplasmic domain

Protein ID accession no.

UniProt accession no.

Gene Information

human ... NLGN4Y(22829)

Specificity

This rabbit polyclonal antibody detects Neuroligin-4. It targets an epitope within 16 amino acids from the C-terminal, cytoplasmic domain.

Immunogen

KLH-conjugated linear peptide corresponding to 16 amino acids from the C-terminal, cytoplasmic domain of human Neuroligin-4.

Application

Quality Control Testing

Evaluated by Western Blotting in lysate from HEK293T cells overexpressing Human Neuroligin-4.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Neuroligin-4 in lysate from HEK293T cells overexpressing Human Neuroligin-4.

Tested Applications

Immunofluorescence Analysis: A representative lot detected Neuroligin-4 in Immunofluorescence applications (Nguyen, T.A., et. al. (2020). Neuron. 106(5):759-768).

Western Blotting Analysis: A representative lot detected Neuroligin-4 in Western Blotting applications (Nguyen, T.A., et. al. (2020). Neuron. 106(5):759-768).

Immunoprecipitation Analysis: A representative lot immunoprecipitated Neuroligin-4 in Immunoprecipitation applications (Nguyen, T.A., et. al. (2020). Neuron. 106(5):759-768).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Neuroligin-4 (UniProt: Q8NFZ3; also known as Neuroligin Y) is encoded by the NLGN4Y (also known as KIAA0951) gene (Gene ID: 22829) in human. Neuroligins are postsynaptic cell adhesion molecules that have crucial roles in synapse maturation and function. In humans, NLGN4X is located on the X chromosome and the NLGN4Y is localized on the Y chromosome and together they form an X-Y pair. They share about 97% amino acid identity; however, they are differentially regulated. Although several mutations in NLGN4X have been linked to autism, only one mutation has been identified in NLGN4Y gene. Despite their high degree of sequence identity, NLGN4Y exhibits severe deficit in trafficking to cell surface and related synaptic effects. This has been attributed to serine 93 in NLGN4Y. A substitution of serine to proline at residue 93 is shown to rescue it and increase its surface trafficking. Neuroligin-4 is a single-pass type I, homodimeric, membrane glycoprotein that is synthesized with a signal peptide (aa 1-43), which is subsequently cleaved off to produce the mature form that contains an extracellular domain (aa 44-676), a transmembrane domain (aa 677-697), and a cytoplasmic domain (aa 698-816). Its expression is observed in fetal and adult brain, prostate, and testis. It serves as a neuronal cell surface protein that is involved in cell-cell interactions. (Ref.: Nguyen, TA., et al. (2020). Neuron 106(5); 759-768; Marro, SG., et al. (2019). Neuron. 103(4); 617-626).

Physical form

Purified rabbit polyclonal antibody in buffer containing PBS, pH 7.4, without azide.

Reconstitution

1.0 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service